Assay Details
Gene Symbol: CHST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005618994.3 | XP_005619051.1 | 3 | 1392 | 67 | |
XM_005618996.3 | XP_005619053.1 | 3 | 1303 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
