Assay Details
Gene Symbol: PABPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130437.2 | NP_001123909.2 | 1 - 2 | 357 | 79 | |
GenBank mRNA | DN272473.1 | - | 1 - 2 | 357 | 79 | |
DN380583.1 | - | 1 - 2 | 241 | 79 | ||
DN400224.1 | - | 1 - 2 | 346 | 79 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
