Assay Details
Gene Symbol: CDYL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005640004.2 | XP_005640061.1 | 7 - 8 | 1695 | 94 | |
XM_005640005.3 | XP_005640062.1 | 5 - 6 | 1455 | 94 | ||
XM_005640006.2 | XP_005640063.1 | 8 - 9 | 1479 | 94 | ||
XM_005640007.2 | XP_005640064.1 | 7 - 8 | 1419 | 94 | ||
XM_022414642.1 | XP_022270350.1 | 5 - 6 | 1338 | 94 | ||
XM_843680.4 | XP_848773.1 | 8 - 9 | 1755 | 94 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
