Assay Details
| Gene Symbol: KCNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003329.1 | NP_001003329.1 | 1 | 975 | 65 | |
| XM_005621307.3 | XP_005621364.1 | 3 | 1666 | 65 | ||
| XM_005621308.3 | XP_005621365.1 | 3 | 1700 | 65 | ||
| GenBank mRNA | L19740.1 | - | 1 | 975 | 65 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

KCNA2
Genomic Map