Assay Details
| Gene Symbol: KCNC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003113.1 | NP_001003113.1 | 1 - 2 | 570 | 90 | |
| XM_022407415.1 | XP_022263123.1 | 1 - 2 | 1030 | 90 | ||
| XM_022407416.1 | XP_022263124.1 | 1 - 2 | 1031 | 90 | ||
| XM_022407417.1 | XP_022263125.1 | 1 - 2 | 1031 | 90 | ||
| GenBank mRNA | AF153198.1 | - | 1 - 2 | 570 | 90 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

KCNC1
Genomic Map