Assay Details
Gene Symbol: PAPSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014110041.2 | XP_013965516.1 | 10 - 11 | 1597 | 107 | |
XM_845977.5 | XP_851070.2 | 11 - 12 | 1708 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
