Assay Details
Gene Symbol: PRICKLE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014111809.2 | XP_013967284.1 | 5 - 6 | 795 | 167 | |
XM_014111810.2 | XP_013967285.1 | 4 - 5 | 648 | 167 | ||
XM_022415809.1 | XP_022271517.1 | 5 - 6 | 661 | 167 | ||
XM_846445.5 | XP_851538.1 | 4 - 5 | 539 | 167 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
