Assay Details
| Gene Symbol: NFE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005636743.3 | XP_005636800.1 | 3 | 667 | 94 | |
| XM_005636744.3 | XP_005636801.1 | 3 | 2258 | 94 | ||
| XM_005636745.3 | XP_005636802.1 | 3 | 1137 | 94 | ||
| XM_005636746.3 | XP_005636803.1 | 3 | 797 | 94 | ||
| XM_543621.5 | XP_543621.2 | 3 | 820 | 94 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NFE2
Genomic Map