Assay Details
Gene Symbol: NKX6-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_544960.6 | XP_544960.2 | 2 - 3 | 2083 | 170 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
