Assay Details
| Gene Symbol: RBM14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014120997.2 | XP_013976472.1 | 1 | 296 | 135 | |
| XM_022405614.1 | XP_022261322.1 | 1 | 298 | 135 | ||
| XM_847190.5 | XP_852283.2 | 1 | 282 | 135 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RBM14
Genomic Map