Assay Details
Gene Symbol: SEC61B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003326.1 | NP_001003326.1 | 2 | 157 | 109 | |
XM_022424897.1 | XP_022280605.1 | 2 | 381 | 109 | ||
GenBank mRNA | L25052.1 | - | 2 | 157 | 109 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
