Assay Details
| Gene Symbol: ST3GAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014114023.2 | XP_013969498.1 | 2 - 3 | 1550 | 156 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ST3GAL2| Gene Symbol: ST3GAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014114023.2 | XP_013969498.1 | 2 - 3 | 1550 | 156 | |
Set Membership: |
|