Assay Details
| Gene Symbol: ARFIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005633608.3 | XP_005633665.1 | 7 - 8 | 1063 | 73 | |
| XM_014122802.1 | XP_013978277.1 | 4 - 5 | 616 | 73 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ARFIP2
Genomic Map