Assay Details
Gene Symbol: PURG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005629887.3 | XP_005629944.1 | 1106 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
