Assay Details
Gene Symbol: REPIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003432079.4 | XP_003432127.1 | 758 | 170 | ||
XM_005629648.3 | XP_005629705.1 | 1181 | 170 | |||
XM_005629649.3 | XP_005629706.1 | 1185 | 170 | |||
XM_005629650.2 | XP_005629707.1 | 684 | 170 | |||
XM_005629651.3 | XP_005629708.1 | 1051 | 170 | |||
XM_005629653.2 | XP_005629710.1 | 498 | 170 | |||
XM_014119877.2 | XP_013975352.1 | 757 | 170 | |||
XM_014119878.2 | XP_013975353.1 | 755 | 170 | |||
XM_022403835.1 | XP_022259543.1 | 1181 | 170 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
