Assay Details
| Gene Symbol: SLC7A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301862.1 | NP_001288791.1 | 1185 | 115 | ||
| XM_540941.6 | XP_540941.3 | 1243 | 115 | |||
| GenBank mRNA | AB847157.1 | - | 1185 | 115 | ||
| EF143580.1 | - | 1057 | 115 | |||
More Information
Set Membership: |
|

SLC7A11
Genomic Map