Assay Details
| Gene Symbol: MTREX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022409389.1 | XP_022265097.1 | 20 - 21 | 2445 | 96 | |
| XM_844583.5 | XP_849676.1 | 19 - 20 | 2381 | 96 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MTREX
Genomic Map