Assay Details
Gene Symbol: GEMIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005624913.3 | XP_005624970.1 | 2 | 2815 | 157 | |
XM_005624915.3 | XP_005624972.1 | 2 | 2823 | 157 | ||
XM_848829.5 | XP_853922.2 | 2 | 2833 | 157 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
