Assay Details
| Gene Symbol: TXNDC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022427913.1 | XP_022283621.1 | 4 - 5 | 348 | 183 | |
| XM_847187.3 | XP_852280.1 | 4 - 5 | 402 | 183 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TXNDC12
Genomic Map