Assay Details
| Gene Symbol: SBDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014114149.1 | XP_013969624.1 | 674 | 170 | ||
| XM_014114150.2 | XP_013969625.1 | 540 | 170 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SBDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014114149.1 | XP_013969624.1 | 674 | 170 | ||
| XM_014114150.2 | XP_013969625.1 | 540 | 170 | |||
Set Membership: |
|