Assay Details
Gene Symbol: ZNF706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014118551.2 | XP_013974026.1 | 391 | 112 | ||
XM_022426589.1 | XP_022282297.1 | 159 | 112 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
