Assay Details
| Gene Symbol: ANAPC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014108057.2 | XP_013963532.1 | 346 | 136 | ||
| XM_022410286.1 | XP_022265994.1 | 214 | 136 | |||
| XM_543390.3 | XP_543390.4 | 382 | 136 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014108057.2 transcripts.
Set Membership: |
|

ANAPC7
Genomic Map