Assay Details
| Gene Symbol: RNF138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022421500.1 | XP_022277208.1 | 6 - 7 | 526 | 75 | |
| XM_022421501.1 | XP_022277209.1 | 6 - 7 | 635 | 75 | ||
| XM_022421502.1 | XP_022277210.1 | 5 - 6 | 463 | 75 | ||
| XM_022421503.1 | XP_022277211.1 | 6 - 7 | 524 | 75 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RNF138
Genomic Map