Assay Details
Gene Symbol: FZD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014107594.2 | XP_013963069.1 | 2 - 3 | 822 | 67 | |
XM_022409709.1 | XP_022265417.1 | 2 - 3 | 701 | 67 | ||
XM_022409710.1 | XP_022265418.1 | 2 - 3 | 820 | 67 | ||
XM_543219.6 | XP_543219.2 | 2 - 3 | 820 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
