Assay Details
Gene Symbol: MED18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005617707.3 | XP_005617764.1 | 3 | 781 | 133 | |
XM_544460.6 | XP_544460.1 | 3 | 794 | 133 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



