Assay Details
| Gene Symbol: PIGV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014110288.2 | XP_013965763.1 | 4 | 804 | 123 | |
| XM_014110291.2 | XP_013965766.1 | 4 | 926 | 123 | ||
| XM_014110292.2 | XP_013965767.1 | 4 | 566 | 123 | ||
| XM_014110295.2 | XP_013965770.1 | 4 | 498 | 123 | ||
| XM_014110298.2 | XP_013965773.1 | 3 | 1250 | 123 | ||
| XM_022412777.1 | XP_022268485.1 | 4 | 353 | 123 | ||
More Information
Set Membership: |
|

PIGV
Genomic Map