Assay Details
| Gene Symbol: PAK1IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022414467.1 | XP_022270175.1 | 6 - 7 | 693 | 81 | |
| XM_535884.6 | XP_535884.1 | 6 - 7 | 718 | 81 | ||
More Information
Set Membership: |
|

PAK1IP1
Genomic Map