Assay Details
Gene Symbol: MED9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_846808.4 | XP_851901.1 | 2 | 441 | 174 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



