Assay Details
| Gene Symbol: SMPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022419292.1 | XP_022275000.1 | 3 - 4 | 1681 | 107 | |
| XM_546863.4 | XP_546863.1 | 3 - 4 | 1683 | 107 | ||
More Information
Set Membership: |
|

SMPD3
Genomic Map