Assay Details
| Gene Symbol: TUBA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022410870.1 | XP_022266578.1 | 2 - 3 | 312 | 184 | |
| XM_022410871.1 | XP_022266579.1 | 4 - 5 | 630 | 184 | ||
| XM_543889.5 | XP_543889.2 | 3 - 4 | 506 | 184 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TUBA8
Genomic Map