Assay Details
| Gene Symbol: GPR85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628745.3 | XP_005628802.1 | 2 | 1437 | 74 | |
| XM_539527.6 | XP_539527.2 | 3 | 1557 | 74 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: GPR85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628745.3 | XP_005628802.1 | 2 | 1437 | 74 | |
| XM_539527.6 | XP_539527.2 | 3 | 1557 | 74 | ||
Set Membership: |
|