Assay Details
| Gene Symbol: CABP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616376.3 | XP_005616433.1 | 1 - 2 | 182 | 166 | |
| XM_541529.6 | XP_541529.2 | 1 - 2 | 66 | 166 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CABP5
Genomic Map