Assay Details
| Gene Symbol: POLE4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022404774.1 | XP_022260482.1 | 1 - 2 | 270 | 69 | |
| XM_022404775.1 | XP_022260483.1 | 1 - 2 | 270 | 69 | ||
| XM_540212.5 | XP_540212.3 | 1 - 2 | 270 | 69 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

POLE4
Genomic Map