Assay Details
Gene Symbol: PMEPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005635212.3 | XP_005635269.1 | 3 - 4 | 406 | 176 | |
XM_005635213.3 | XP_005635270.1 | 3 - 4 | 2509 | 176 | ||
XM_005635214.3 | XP_005635271.1 | 3 - 4 | 2506 | 176 | ||
XM_005635215.3 | XP_005635272.1 | 3 - 4 | 411 | 176 | ||
XM_022408861.1 | XP_022264569.1 | 3 - 4 | 403 | 176 | ||
XM_022408862.1 | XP_022264570.1 | 3 - 4 | 418 | 176 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
