Assay Details
| Gene Symbol: RGMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005618321.3 | XP_005618378.2 | 3 - 4 | 731 | 118 | |
| XM_014112400.2 | XP_013967875.1 | 3 - 4 | 3845 | 118 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RGMA
Genomic Map