Assay Details
| Gene Symbol: KCTD16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617336.3 | XP_005617393.1 | 1384 | 154 | ||
| XM_005617337.3 | XP_005617394.1 | 1188 | 154 | |||
| XM_014108141.2 | XP_013963616.1 | 1265 | 154 | |||
More Information
Set Membership: |
|

KCTD16
Genomic Map