Assay Details
| Gene Symbol: MCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003016.2 | NP_001003016.2 | 2 - 3 | 1014 | 116 | |
| GenBank mRNA | AB093582.2 | - | 2 - 3 | 1041 | 116 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MCL1
Genomic Map