Assay Details
| Gene Symbol: GFRA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022408943.1 | XP_022264651.1 | 3 | 1429 | 122 | |
| XM_022408944.1 | XP_022264652.1 | 3 | 1532 | 148 | ||
More Information
Set Membership: |
|

GFRA4
Genomic Map