Assay Details
| Gene Symbol: GRAMD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005626468.3 | XP_005626525.1 | 8 - 9 | 909 | 147 | |
| XM_531891.6 | XP_531891.3 | 8 - 9 | 849 | 147 | ||
More Information
Set Membership: |
|

GRAMD2B
Genomic Map