Assay Details
| Gene Symbol: TRAPPC6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616452.3 | XP_005616509.2 | 4 - 5 | 427 | 147 | |
| XM_005616453.3 | XP_005616510.2 | 4 - 5 | 361 | 147 | ||
| XM_014119739.2 | XP_013975214.2 | 3 - 4 | 344 | 147 | ||
More Information
Set Membership: |
|

TRAPPC6A
Genomic Map