Assay Details
Gene Symbol: GEMIN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003432193.3 | XP_003432241.1 | 3 | 442 | 184 | |
XM_022404652.1 | XP_022260360.1 | 4 | 1161 | 184 | ||
XM_532941.5 | XP_532941.2 | 3 | 256 | 184 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
