Assay Details
Gene Symbol: RAI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014113665.2 | XP_013969140.1 | 2 | 5173 | 125 | |
XM_022418657.1 | XP_022274365.1 | 3 | 5052 | 125 | ||
XM_022418658.1 | XP_022274366.1 | 2 | 5572 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
