Assay Details
| Gene Symbol: RSPH6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022424823.1 | XP_022280531.1 | 3 - 4 | 1862 | 125 | |
| XM_533639.6 | XP_533639.1 | 3 - 4 | 1862 | 125 | ||
More Information
Set Membership: |
|

RSPH6A
Genomic Map