Assay Details
| Gene Symbol: HM13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022408849.1 | XP_022264557.1 | 7 - 8 | 946 | 67 | |
| XM_845723.5 | XP_850816.1 | 6 - 7 | 761 | 67 | ||
| XM_857030.6 | XP_862123.1 | 6 - 7 | 759 | 67 | ||
More Information
Set Membership: |
|

HM13
Genomic Map