Assay Details
Gene Symbol: CPLANE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005617960.2 | XP_005618017.1 | 4 | 510 | 98 | |
XM_005617961.3 | XP_005618018.1 | 4 | 521 | 98 | ||
XM_544544.6 | XP_544544.3 | 4 | 605 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
