Assay Details
| Gene Symbol: HDHD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005626944.2 | XP_005627001.1 | 2 | 549 | 161 | |
| XM_022424961.1 | XP_022280669.1 | 2 | 700 | 161 | ||
More Information
Set Membership: |
|

HDHD3
Genomic Map