Assay Details
Gene Symbol: PROK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639116.3 | XP_003639164.1 | 2 - 3 | 389 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
