Assay Details
Gene Symbol: SPSB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637212.3 | XP_005637269.1 | 2 | 862 | 89 | |
XM_005637213.3 | XP_005637270.1 | 2 | 867 | 89 | ||
XM_005637214.3 | XP_005637271.1 | 2 | 802 | 89 | ||
XM_022410961.1 | XP_022266669.1 | 2 | 754 | 89 | ||
XM_022410962.1 | XP_022266670.1 | 2 | 978 | 89 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
