Assay Details
| Gene Symbol: SEC11C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003312.1 | NP_001003312.1 | 732 | 150 | ||
| GenBank mRNA | J05069.1 | - | 732 | 150 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SEC11C
Genomic Map