Assay Details
Gene Symbol: LRRC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005634346.3 | XP_005634403.1 | 2 | 424 | 147 | |
XM_005634348.3 | XP_005634405.1 | 2 | 496 | 147 | ||
XM_005634349.3 | XP_005634406.1 | 2 | 512 | 147 | ||
XM_014106810.2 | XP_013962285.1 | 2 | 515 | 147 | ||
XM_014106811.2 | XP_013962286.1 | 2 | 508 | 147 | ||
XM_022408564.1 | XP_022264272.1 | 3 | 660 | 147 | ||
XM_542758.7 | XP_542758.1 | 2 | 506 | 147 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
